missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-[3-(chloromethyl)phenyl]pyrimidine, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5413506

 View more versions of this product

Catalog No. AAH5413506



Specifications

Specifications

5-Bromo-2-[3-(chloromethyl)phenyl]pyrimidine
1100598-50-2
MFCD20527211
KHSZEUJOVUJWKR-UHFFFAOYSA-N
5-bromo-2-[3-(chloromethyl)phenyl]pyrimidine
59438047
97%
5g
C11H8BrClN2
5-bromo-2-3-chloromethyl phenyl pyrimidine
C1=CC(=CC(=C1)CCl)C2=NC=C(C=N2)Br
283.553
283.55

Chemical Identifiers

1100598-50-2
283.553
KHSZEUJOVUJWKR-UHFFFAOYSA-N
59438047
C1=CC(=CC(=C1)CCl)C2=NC=C(C=N2)Br
C11H8BrClN2
MFCD20527211
5-bromo-2-3-chloromethyl phenyl pyrimidine
5-bromo-2-[3-(chloromethyl)phenyl]pyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents