missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-(2-thienyl)pyridine, 95%

$167.78 - $167.78

Chemical Identifiers

CAS 91891-74-6
Molecular Formula C9H6BrNS
Formula Weight 240.12
MDL Number MFCD09037419
Chemical Name or Material 5-Bromo-2-(2-thienyl)pyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH27237MD SDS Alfa Aesar™
H27237MD
250mg Each for $167.78
AAH2723703 SDS Alfa Aesar™
H2723703
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

91891-74-6
240.12
5-Bromo-2-(2-thienyl)pyridine
C9H6BrNS
MFCD09037419

Specifications

91891-74-6
240.12
250mg
95%
C9H6BrNS
MFCD09037419
5-Bromo-2-(2-thienyl)pyridine

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger