missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-(2-furyl)pyridine, 95%

$227.14 - $513.04

Chemical Identifiers

CAS 942206-06-6
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD09037418
InChI Key ZKUDFBZRIQOMGO-UHFFFAOYSA-N
Synonym 5-bromo-2-2-furyl pyridine, 5-bromo-2-fur-2-yl pyridine, 5-bromo-2-furan-2-yl pyridine
PubChem CID 42552976
IUPAC Name 5-bromo-2-(furan-2-yl)pyridine
SMILES C1=COC(=C1)C2=NC=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH26942MD
View Documents
Alfa Aesar™
H26942MD
250mg
Each for $227.14
 
AAH2694203
View Documents
Alfa Aesar™
H2694203
1g
Each for $513.04
 
Description

Description

Specifications

Chemical Identifiers

942206-06-6
224.057
ZKUDFBZRIQOMGO-UHFFFAOYSA-N
42552976
C1=COC(=C1)C2=NC=C(C=C2)Br
C9H6BrNO
MFCD09037418
5-bromo-2-2-furyl pyridine, 5-bromo-2-fur-2-yl pyridine, 5-bromo-2-furan-2-yl pyridine
5-bromo-2-(furan-2-yl)pyridine

Specifications

250mg
C9H6BrNO
5-bromo-2-2-furyl pyridine, 5-bromo-2-fur-2-yl pyridine, 5-bromo-2-furan-2-yl pyridine
C1=COC(=C1)C2=NC=C(C=C2)Br
224.057
224.06
5-Bromo-2-(2-furyl)pyridine
942206-06-6
MFCD09037418
ZKUDFBZRIQOMGO-UHFFFAOYSA-N
5-bromo-2-(furan-2-yl)pyridine
42552976
95%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS