Please login to your online account to display your discounted pricing

5-bromo-1h-indazole, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Bromo-1H-indazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H5BrN2
Formula Weight 197.03
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

CC48210CB

  SDS
fisher scientific company
CC48210CB
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC48210DA

  SDS
fisher scientific company
CC48210DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.03
Quantity 250mg
Assay 97%
Chemical Name or Material 5-Bromo-1H-indazole
Assay Percent Range 97%
Molecular Formula C7H5BrN2
CAS 53857-57-1
Health Hazard 2 GHS P Statement May cause respiratory irritation. Toxic if swallowed. Causes serious eye irritation. Causes skin irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement May cause respiratory irritation. Toxic if swallowed. Causes serious eye irritation. Causes skin irritation.