Please login to your online account to display your discounted pricing

5-Bromo-1H-benzimidazole, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC45910EA

 View more versions of this product

Catalog No. CC45910EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 197.03
Quantity 10g
Assay 97%
CAS 4887-88-1
Chemical Name or Material 5-Bromo-1H-benzimidazole
Packaging Amber Glass Bottle
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Avoid contact during pregnancy/while nursing.
Health Hazard 2 GHS P Statement Harmful if swallowed.
Molecular Formula C7H5BrN2
Assay Percent Range 97%

Specifications

Chemical Name or Material 5-Bromo-1H-benzimidazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H5BrN2
Formula Weight 197.03
Accessories

Warning

GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Avoid contact during pregnancy/while nursing.

GHS P Statement Harmful if swallowed.