Please login to your online account to display your discounted pricing

5-Bromo-1-(triisopropylsilyl)-1H-indole, ≥97%, Maybridge

$153.70 - $153.70

Chemical Identifiers

Chemical Name or Material 5-Bromo-1-(triisopropylsilyl)-1H-indole
CAS 128564-66-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C17H26BrNSi
Formula Weight 352.39
MDL Number MFCD05664417
Synonym 5-Bromo-1-(triisopropylsilyl)-1H-indole (IUPAC)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00980DA

 
fisher scientific company
MO00980DA
Amber Glass Bottle 1g Each for $153.70

MO00980EA

 
fisher scientific company
MO00980EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 352.39
Quantity 1g
Assay 97%
CAS Min % 97.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 5-Bromo-1-(triisopropylsilyl)-1H-indole
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C17H26BrNSi
MDL Number MFCD05664417
CAS 128564-66-9
Synonym 5-Bromo-1-(triisopropylsilyl)-1H-indole (IUPAC)
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.