Please login to your online account to display your discounted pricing

5-Bromo-1-(tert-butoxycarbonyl)-1H-indol-2-ylboronic acid, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Bromo-1-(tert-butoxycarbonyl)-1H-indol-2-ylboronic acid
CAS 475102-13-7
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C13H15BBrNO4
Formula Weight 339.98
MDL Number MFCD03701683
Synonym 5-Bromo-1-(tert-butoxycarbonyl)-1H-indol-2-ylboronic acid (IUPAC)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO01243DE

 
fisher scientific company
MO01243DE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 339.98
Quantity 5g
Assay 95%
CAS Min % 95.0
MDL Number MFCD03701683
Assay Percent Range 95%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym 5-Bromo-1-(tert-butoxycarbonyl)-1H-indol-2-ylboronic acid (IUPAC)
Chemical Name or Material 5-Bromo-1-(tert-butoxycarbonyl)-1H-indol-2-ylboronic acid
CAS 475102-13-7
Molecular Formula C13H15BBrNO4
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.