missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-1-methyl-2-oxoindoline, 97%, Maybridge

$124.16 - $124.16

Chemical Identifiers

CAS 20870-90-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H8BrNO
Formula Weight 226.07
MDL Number MFCD00464061
Chemical Name or Material 5-Bromo-1-methyl-2-oxoindoline
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC63010DA SDS Fisher Scientific Company
CC63010DA
Amber Glass Bottle 1g Each for $124.16
CC63010DE SDS Fisher Scientific Company
CC63010DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC63010EA SDS Fisher Scientific Company
CC63010EA
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

20870-90-0
100.0
C9H8BrNO
MFCD00464061
97%
96.5
97%
226.07
5-Bromo-1-methyl-2-oxoindoline

Specifications

226.07
100.0
MFCD00464061
Amber Glass Bottle
C9H8BrNO
97%
1g
97%
96.5
20870-90-0
5-Bromo-1-methyl-2-oxoindoline

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning