Please login to your online account to display your discounted pricing

5-bromo-1-methyl-1H-indole-3-carbaldehyde, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC75304DA

 View more versions of this product

Catalog No. CC75304DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 238.08
Quantity 1g
Assay 97%
Molecular Formula C10H8BrNO
CAS 10102-94-0
Packaging Amber Glass Bottle
Chemical Name or Material 5-bromo-1-methyl-1H-indole-3-carbaldehyde

Specifications

Chemical Name or Material 5-bromo-1-methyl-1H-indole-3-carbaldehyde
Assay 97%
Molecular Formula C10H8BrNO
Formula Weight 238.08