Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-1-isoindolinone, 96%

$200.11 - $200.11

Chemical Identifiers

Chemical Name or Material 5-Bromo-1-isoindolinone
CAS 552330-86-6
Molecular Formula C8H6BrNO
Formula Weight 212.04
MDL Number MFCD09701292
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH52358MD

 
alfa aesar™
H52358MD
250mg Each for $200.11
Description & Specifications

Specifications

Formula Weight 212.04
Chemical Name or Material 5-Bromo-1-isoindolinone
Assay 96%
Molecular Formula C8H6BrNO
MDL Number MFCD09701292
CAS 552330-86-6
Quantity 250mg