Please login to your online account to display your discounted pricing

5-Bromo-1-benzofuran, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC07110DE

 View more versions of this product

Catalog No. CC07110DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 197.03
Quantity 5g
Assay 97%
CAS Max % 100.0
Health Hazard 1 Warning
Chemical Name or Material 5-Bromo-1-benzofuran
Molecular Formula C8H5BrO
MDL Number MFCD03407317
CAS 23145-07-5
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Assay Percent Range 97%
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-Bromo-1-benzofuran
CAS 23145-07-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H5BrO
Formula Weight 197.03
MDL Number MFCD03407317
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.