Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-1,2,4-trimethylbenzene, 99%

$40.69 - $97.64

Chemical Identifiers

Chemical Name or Material 5-Bromo-1,2,4-trimethylbenzene
CAS 5469-19-2
Molecular Formula C9H11Br
Formula Weight 199.09
MDL Number MFCD00000072
Synonym 2,4,5-Trimethylbromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2327114

  SDS
alfa aesar™
B2327114
25g Each for $40.69

AAB2327122

  SDS
alfa aesar™
B2327122
100g Each for $97.64
Description & Specifications

Specifications

Formula Weight 199.09
Assay 99%
Chemical Name or Material 5-Bromo-1,2,4-trimethylbenzene
CAS 5469-19-2
Boiling Point 233° to 235°C
Synonym 2,4,5-Trimethylbromobenzene
Melting Point 71° to 73°C
MDL Number MFCD00000072
Molecular Formula C9H11Br
Quantity 25g