Learn More
5-Bromo-1,2,3-trimethoxybenzene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2487206
Description
5-Bromo-1,2,3-trimethoxybenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromo-1,2,3-trimethoxybenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
5-Bromo-1,2,3-trimethoxybenzene | |
77°C to 80°C | |
MFCD00017169 | |
1-bromo-3,4,5-trimethoxybenzene, 3,4,5-trimethoxybromobenzene, bromotrimethoxybenzene, benzene, 5-bromo-1,2,3-trimethoxy, 5-bromo-1,2,3-trimethoxy-benzene, pubchem10980, 3,4,5-trimethoxybromoben, ksc490o1d, 3,4,5-trimethoxyphenylbromide, 3,4,5-triemethoxyphenylbromide | |
XAOOZMATJDXDQJ-UHFFFAOYSA-N | |
5-bromo-1,2,3-trimethoxybenzene | |
75885 | |
97% |
2675-79-8 | |
C9H11BrO3 | |
5 g | |
Insoluble in water. | |
COC1=CC(Br)=CC(OC)=C1OC | |
247.09 | |
247.1 |
Chemical Identifiers
2675-79-8 | |
247.09 | |
XAOOZMATJDXDQJ-UHFFFAOYSA-N | |
75885 | |
COC1=CC(Br)=CC(OC)=C1OC |
C9H11BrO3 | |
MFCD00017169 | |
1-bromo-3,4,5-trimethoxybenzene, 3,4,5-trimethoxybromobenzene, bromotrimethoxybenzene, benzene, 5-bromo-1,2,3-trimethoxy, 5-bromo-1,2,3-trimethoxy-benzene, pubchem10980, 3,4,5-trimethoxybromoben, ksc490o1d, 3,4,5-trimethoxyphenylbromide, 3,4,5-triemethoxyphenylbromide | |
5-bromo-1,2,3-trimethoxybenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
EINECSNumber : 220-223-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only