Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-1,2,3-trifluorobenzene, 98%

Chemical Identifiers

Chemical Name or Material 5-Bromo-1,2,3-trifluorobenzene
CAS 138526-69-9
Molecular Formula C6H2BrF3
Formula Weight 210.98
MDL Number MFCD00070738
UN Number UN1993
Beilstein 7249191
Synonym 1-Bromo-3,4,5-trifluorobenzene; 3,4,5-Trifluorobromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2060703

  SDS
alfa aesar™
B2060703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2060706

  SDS
alfa aesar™
B2060706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.98
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Assay 98%
Health Hazard 2 GHS H Statement
H318-H226-H351-H315
Causes serious eye damage.
Flammable liquid and vapour.
Suspected of causing cancer.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 7249191
UN Number UN1993
Chemical Name or Material 5-Bromo-1,2,3-trifluorobenzene
Melting Point <−20°C
Synonym 1-Bromo-3,4,5-trifluorobenzene; 3,4,5-Trifluorobromobenzene
Flash Point 45°C (113°F)
Quantity 1g
CAS 138526-69-9
MDL Number MFCD00070738
Molecular Formula C6H2BrF3
Boiling Point 140°C
Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H226-H351-H315
Causes serious eye damage.
Flammable liquid and vapour.
Suspected of causing cancer.
Causes skin irritation.