missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(Boc-amino)valeric acid, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6616803

 View more versions of this product

Catalog No. AAH6616803



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

5-(Boc-amino)valeric acid
48°C to 52°C
160°C to 168°C (0.8 mmHg)
C10H19NO4
MFCD00076903
2048862
GFMRZAMDGJIWRB-UHFFFAOYSA-N
5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
545848
95%
27219-07-4
White
113°C (235°F)
(CH3)3COC(=O)NH(CH2)4CO2H
1 g
boc-5-ava-oh, 5-tert-butoxycarbonylamino valeric acid, boc-5-aminovaleric acid, 5-boc-amino valeric acid, 5-tert-butoxycarbonyl amino pentanoic acid, 5-boc-amino-pentanoic acid, n-boc-5-aminopentanoic acid, pentanoic acid, 5-1,1-dimethylethoxy carbonyl amino, n-boc-5-aminovaleric acid
CC(C)(C)OC(=O)NCCCCC(=O)O
217.265
217.27
Powder

Chemical Identifiers

27219-07-4
217.265
GFMRZAMDGJIWRB-UHFFFAOYSA-N
545848
CC(C)(C)OC(=O)NCCCCC(=O)O
C10H19NO4
MFCD00076903
boc-5-ava-oh, 5-tert-butoxycarbonylamino valeric acid, boc-5-aminovaleric acid, 5-boc-amino valeric acid, 5-tert-butoxycarbonyl amino pentanoic acid, 5-boc-amino-pentanoic acid, n-boc-5-aminopentanoic acid, pentanoic acid, 5-1,1-dimethylethoxy carbonyl amino, n-boc-5-aminovaleric acid
5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions