Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(Boc-amino)pyridine-3-boronic acid pinacol ester, 95%

Chemical Identifiers

Chemical Name or Material 5-(Boc-amino)pyridine-3-boronic acid pinacol ester
CAS 1171897-39-4
Formula Weight 320.2
MDL Number MFCD13195277
Beilstein 19330771
Synonym [5-(4,4,5,5-Tetramethyl-1,3,2dioxaborolan-2-yl)-3-pyridyl]carbamic acid tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66223MD

 
alfa aesar™
H66223MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 320.2
Physical Form Solid
Assay 95%
MDL Number MFCD13195277
CAS 1171897-39-4
Quantity 250mg
Beilstein 19330771
Synonym [5-(4,4,5,5-Tetramethyl-1,3,2dioxaborolan-2-yl)-3-pyridyl]carbamic acid tert-butyl ester
Chemical Name or Material 5-(Boc-amino)pyridine-3-boronic acid pinacol ester