Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(beta-Styryl)-2H-1,2,3,4-tetrazole, 99%

$708.18 - $708.18

Chemical Identifiers

Chemical Name or Material 5-(beta-Styryl)-2H-1,2,3,4-tetrazole
CAS 220429-71-0
Molecular Formula C9H8N4
Formula Weight 172.19
MDL Number MFCD00276928
Beilstein 6502443
Synonym 5-(2-Phenylethenyl)-2H-tetraazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5110706

  SDS
alfa aesar™
H5110706
5g Each for $708.18
Description & Specifications

Specifications

Formula Weight 172.19
Chemical Name or Material 5-(beta-Styryl)-2H-1,2,3,4-tetrazole
Assay 99%
Molecular Formula C9H8N4
MDL Number MFCD00276928
CAS 220429-71-0
Beilstein 6502443
Synonym 5-(2-Phenylethenyl)-2H-tetraazole
Quantity 5g