Learn More
5-Benzyloxytryptamine hydrochloride, 98%, Thermo Scientific Chemicals
$306.57 - $306.57
Chemical Identifiers
CAS | 52055-23-9 |
---|---|
Molecular Formula | C17H18N2O·HCl |
Molecular Weight (g/mol) | 302.8 |
MDL Number | MFCD00012685 |
InChI Key | IUWVJCIEWSQGHH-UHFFFAOYSA-N |
Synonym | 5-benzyloxytryptamine hydrochloride, 5-benzyloxytryptamine hcl, 2-5-phenylmethoxy-1h-indol-3-yl ethanamine hydrochloride, cambridge id 5119598, c17h18n2o.hcl, 5-benzyloxy-tryptamine hydrochloride, 5-benzyloxy tryptamine hydrochloride, 5-benzyloxytryptamine hydrochloridyl, 3-2-aminoethyl-5-benzyloxyindole hydrochloride |
PubChem CID | 2828785 |
IUPAC Name | 2-(5-phenylmethoxy-1H-indol-3-yl)ethanamine;hydrochloride |
SMILES | C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3CCN.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC106110010
|
Thermo Scientific Chemicals
106110010 |
1 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52055-23-9 | |
302.8 | |
IUWVJCIEWSQGHH-UHFFFAOYSA-N | |
2828785 | |
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3CCN.Cl |
C17H18N2O·HCl | |
MFCD00012685 | |
5-benzyloxytryptamine hydrochloride, 5-benzyloxytryptamine hcl, 2-5-phenylmethoxy-1h-indol-3-yl ethanamine hydrochloride, cambridge id 5119598, c17h18n2o.hcl, 5-benzyloxy-tryptamine hydrochloride, 5-benzyloxy tryptamine hydrochloride, 5-benzyloxytryptamine hydrochloridyl, 3-2-aminoethyl-5-benzyloxyindole hydrochloride | |
2-(5-phenylmethoxy-1H-indol-3-yl)ethanamine;hydrochloride |
Specifications
52055-23-9 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00012685 | |
5-benzyloxytryptamine hydrochloride, 5-benzyloxytryptamine hcl, 2-5-phenylmethoxy-1h-indol-3-yl ethanamine hydrochloride, cambridge id 5119598, c17h18n2o.hcl, 5-benzyloxy-tryptamine hydrochloride, 5-benzyloxy tryptamine hydrochloride, 5-benzyloxytryptamine hydrochloridyl, 3-2-aminoethyl-5-benzyloxyindole hydrochloride | |
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3CCN.Cl | |
302.8 | |
302.8 | |
Crystalline Powder |
97.5 | |
255°C to 259°C | |
97.5% min. (HPLC) | |
C17H18N2O·HCl | |
1 g | |
IUWVJCIEWSQGHH-UHFFFAOYSA-N | |
2-(5-phenylmethoxy-1H-indol-3-yl)ethanamine;hydrochloride | |
2828785 | |
98% | |
5-Benzyloxytryptamine hydrochloride, 98% |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 257-626-4
RUO – Research Use Only