Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Benzyloxytryptamine hydrochloride, 98%

$223.00 - $223.00

Chemical Identifiers

Chemical Name or Material 5-Benzyloxytryptamine hydrochloride
CAS 52055-23-9
Molecular Formula C17H18N2O·HCl
Formula Weight 302.8
MDL Number MFCD00012685
Synonym 3-(2-Aminoethyl)-5-benzyloxyindole hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5108903

  SDS
alfa aesar™
H5108903
1g Each for $223.00

AAH51089MD

  SDS
alfa aesar™
H51089MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 5-Benzyloxytryptamine hydrochloride
CAS 52055-23-9
Assay 98%
Molecular Formula C17H18N2O·HCl
Formula Weight 302.8
MDL Number MFCD00012685
Synonym 3-(2-Aminoethyl)-5-benzyloxyindole hydrochloride
Melting Point 259° to 263°C
Quantity 1g