Please login to your online account to display your discounted pricing

5-Benzyloxyindole 95%, ACROS Organics™


Quantity 10g

Manufacturer: thermo fisher scientific  106090100

 View more versions of this product

Catalog No. AC106090100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 223.27
Quantity 10g
Color Off-white to beige
EINECS Number 214-930-1
RTECS Number NL4850000
CAS Min % 94.0
CAS Max % 100.0
Health Hazard 1 Warning
Individual Impurity Toluene: < 6% (GC)
Assay Percent Range 95%
Molecular Formula C15H13NO
CAS 1215-59-4
Chemical Name or Material 5-Benzyloxyindole
Infrared Spectrum Authentic
Name Note 95%
MDL Number MFCD00005676
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-Benzyloxyindole
Name Note 95%
CAS 1215-59-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C15H13NO
Formula Weight 223.27
MDL Number MFCD00005676
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents