Learn More
5-Benzyloxyindole, 94%, may contain up to ca 7% toluene, Thermo Scientific Chemicals
$62.19 - $564.07
Chemical Identifiers
CAS | 1215-59-4 |
---|---|
Molecular Formula | C15H13NO |
Molecular Weight (g/mol) | 223.275 |
MDL Number | MFCD00005676 |
InChI Key | JCQLPDZCNSVBMS-UHFFFAOYSA-N |
Synonym | 5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c |
PubChem CID | 14624 |
IUPAC Name | 5-phenylmethoxy-1H-indole |
SMILES | C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2514303
|
Thermo Scientific Chemicals
B2514303 |
1 g |
Each for $62.19
|
|
|||||
AAB2514306
|
Thermo Scientific Chemicals
B2514306 |
5 g |
Each for $182.42
|
|
|||||
AAB2514314
|
Thermo Scientific Chemicals
B2514314 |
25 g |
Each for $564.07
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1215-59-4 | |
223.275 | |
JCQLPDZCNSVBMS-UHFFFAOYSA-N | |
14624 | |
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3 |
C15H13NO | |
MFCD00005676 | |
5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c | |
5-phenylmethoxy-1H-indole |
Specifications
1215-59-4 | |
C15H13NO | |
1 g | |
5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c | |
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3 | |
223.275 | |
223.28 | |
5-Benzyloxyindole |
96°C to 101°C | |
MFCD00005676 | |
173532 | |
JCQLPDZCNSVBMS-UHFFFAOYSA-N | |
5-phenylmethoxy-1H-indole | |
14624 | |
94% | |
may contain up to ≈7% toluene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P331-P332+P313-P362-P501c
H302+H312+H332-H304-H315-H319-H335-H336-H361d
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 214-930-1
RTECSNumber : NL4850000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only