missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Benzyloxyindole-2-carboxylic acid, 97%

$100.84 - $347.05

Chemical Identifiers

CAS 6640-09-1
Molecular Formula C16H13NO3
Formula Weight 267.29
MDL Number MFCD00047165
Beilstein 241643
Chemical Name or Material 5-Benzyloxyindole-2-carboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0087803 SDS Alfa Aesar™
L0087803
1g Each for $100.84
AAL0087806 SDS Alfa Aesar™
L0087806
5g Each for $347.05
Specifications Description & Specifications

Chemical Identifiers

6640-09-1
267.29
241643
C16H13NO3
MFCD00047165
5-Benzyloxyindole-2-carboxylic acid

Specifications

6640-09-1
267.29
241643
1g
97%
C16H13NO3
MFCD00047165
194° to 200°C
5-Benzyloxyindole-2-carboxylic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning