Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(Benzyloxycarbonylamino)valeric acid, 97%

Chemical Identifiers

Chemical Name or Material 5-(Benzyloxycarbonylamino)valeric acid
CAS 23135-50-4
Linear Formula C6H4CH2OCONH2(CH2)4CO2H
Formula Weight 251.28
MDL Number MFCD00188458
Beilstein 2334766
Synonym 5-(Benzyloxycarbonylamino)pentanoic acid; N-Cbz-5-aminovaleric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6495803

 
alfa aesar™
H6495803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6495806

 
alfa aesar™
H6495806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64958MD

 
alfa aesar™
H64958MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.28
Melting Point 99° to 105°C
Chemical Name or Material 5-(Benzyloxycarbonylamino)valeric acid
Physical Form Solid
Assay 97%
Linear Formula C6H4CH2OCONH2(CH2)4CO2H
MDL Number MFCD00188458
CAS 23135-50-4
Beilstein 2334766
Quantity 1g
Color Off-white
Synonym 5-(Benzyloxycarbonylamino)pentanoic acid; N-Cbz-5-aminovaleric acid