Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Benzyloxy-6-azaindole-3-carboxaldehyde, 96%

$143.45 - $474.64

Chemical Identifiers

Chemical Name or Material 5-Benzyloxy-6-azaindole-3-carboxaldehyde
CAS 56795-92-7
Molecular Formula C15H12N2O2
Formula Weight 252.28
MDL Number MFCD00191784
Synonym 5-Benzyloxy-1H-pyrrolo[2,3-c]pyridine-3-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5017303

  SDS
alfa aesar™
H5017303
1g Each for $474.64

AAH50173MD

  SDS
alfa aesar™
H50173MD
250mg Each for $143.45
Description & Specifications

Specifications

Formula Weight 252.28
Health Hazard 1 Warning
Chemical Name or Material 5-Benzyloxy-6-azaindole-3-carboxaldehyde
Molecular Formula C15H12N2O2
MDL Number MFCD00191784
CAS 56795-92-7
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Assay 96%
Synonym 5-Benzyloxy-1H-pyrrolo[2,3-c]pyridine-3-carboxaldehyde
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.