missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Benzyloxy-3-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6259803

 View more versions of this product

Catalog No. AAH6259803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Benzyloxy-3-fluorobenzeneboronic acid pinacol ester
1528757-88-1
3-Benzyloxy-5-fluorophenylboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)OCC3=CC=CC=C3
328.19
328.19
96%
1g
C19H22BFO3
FAVUCVNUXHFOPD-UHFFFAOYSA-N
2-(3-fluoro-5-phenylmethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
81315028
Solid

Chemical Identifiers

1528757-88-1
328.19
3-Benzyloxy-5-fluorophenylboronic acid pinacol ester
2-(3-fluoro-5-phenylmethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C19H22BFO3
FAVUCVNUXHFOPD-UHFFFAOYSA-N
81315028
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)OCC3=CC=CC=C3
Documents