Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Benzyloxy-3-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 5-Benzyloxy-3-fluorobenzeneboronic acid pinacol ester
Formula Weight 328.19
Synonym 3-Benzyloxy-5-fluorophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6259803

 
alfa aesar™
H6259803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62598MD

 
alfa aesar™
H62598MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 328.19
Chemical Name or Material 5-Benzyloxy-3-fluorobenzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
Quantity 1g
Synonym 3-Benzyloxy-5-fluorophenylboronic acid pinacol ester