Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Benzyloxy-2-nitrotoluene, 98%

$47.32 - $158.22

Chemical Identifiers

Chemical Name or Material 5-Benzyloxy-2-nitrotoluene
CAS 22424-58-4
Molecular Formula C14H13NO3
Formula Weight 243.26
MDL Number MFCD00024269
Beilstein 1978227
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0353803

  SDS
alfa aesar™
L0353803
1g Each for $47.32

AAL0353806

  SDS
alfa aesar™
L0353806
5g Each for $158.22
Description & Specifications

Specifications

Formula Weight 243.26
Chemical Name or Material 5-Benzyloxy-2-nitrotoluene
Molecular Formula C14H13NO3
CAS 22424-58-4
Assay 98%
MDL Number MFCD00024269
Beilstein 1978227
Melting Point 69° to 70°C
Quantity 1g