missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Benzyl N-(tert-Butoxycarbonyl)-L-glutamate, TCI America™

$38.19 - $143.61

Chemical Identifiers

CAS 13574-13-5
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.372
MDL Number MFCD00065569
InChI Key AJDUMMXHVCMISJ-ZDUSSCGKSA-N
Synonym boc-glu obzl-oh, boc-l-glutamic acid 5-benzyl ester, boc-l-glutamic acid 5-benzylester, 5-benzyl n-boc-l-glutamate, boc-l-glu obzl-oh, n-alpha-tert-boc-l-glutamic-gamma-benzyl ester, 5-benzyl n-tert-butoxycarbonyl-l-glutamate, boc-l-glutamic acid-5-benzyl ester, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, 5-phenylmethyl ester, n-boc-l-glutamic acid 5-benzyl ester
PubChem CID 83589
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B16335G View Documents TCI America
B16335G
5g Each for $38.19
 
B163325G View Documents TCI America
B163325G
25g Each for $143.61
 
Specifications

Chemical Identifiers

13574-13-5
337.372
AJDUMMXHVCMISJ-ZDUSSCGKSA-N
83589
CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(=O)O
C17H23NO6
MFCD00065569
boc-glu obzl-oh, boc-l-glutamic acid 5-benzyl ester, boc-l-glutamic acid 5-benzylester, 5-benzyl n-boc-l-glutamate, boc-l-glu obzl-oh, n-alpha-tert-boc-l-glutamic-gamma-benzyl ester, 5-benzyl n-tert-butoxycarbonyl-l-glutamate, boc-l-glutamic acid-5-benzyl ester, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, 5-phenylmethyl ester, n-boc-l-glutamic acid 5-benzyl ester
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid

Specifications

White
5g
C17H23NO6
boc-glu obzl-oh, boc-l-glutamic acid 5-benzyl ester, boc-l-glutamic acid 5-benzylester, 5-benzyl n-boc-l-glutamate, boc-l-glu obzl-oh, n-alpha-tert-boc-l-glutamic-gamma-benzyl ester, 5-benzyl n-tert-butoxycarbonyl-l-glutamate, boc-l-glutamic acid-5-benzyl ester, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, 5-phenylmethyl ester, n-boc-l-glutamic acid 5-benzyl ester
CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(=O)O
337.372
337.37
≥98.0% (HPLC,T)
68°C
13574-13-5
MFCD00065569
AJDUMMXHVCMISJ-ZDUSSCGKSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
83589
Crystal-Powder at 20°C
5-Benzyl N-(tert-Butoxycarbonyl)-L-glutamate
Safety and Handling

Safety and Handling

TSCA : Yes