missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Benzyl N-Carbobenzoxy-L-glutamate 98.0+%, TCI America™

Manufacturer:  TCI America B390225G

 View more versions of this product

Catalog No. B390225G



Specifications

Specifications

5-Benzyl N-Carbobenzoxy-L-glutamate
76°C
5680-86-4
MFCD00065697
TWIVXHQQTRSWGO-KRWDZBQOSA-N
(2S)-5-oxo-5-phenylmethoxy-2-(phenylmethoxycarbonylamino)pentanoic acid
3049860
Crystal-Powder at 20°C
White
25g
C20H21NO6
z-glu obzl-oh, cbz-glu obzl-oh, 2s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, 5-benzyl n-carbobenzoxy-l-glutamate, n-cbz-l-glutamic acid 5-benzyl ester, s-2-benzyloxycarbonylamino-pentanedioic acid 5-benzyl ester, s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, z-glu obzl oh, pubchem14964, z-glu-obzl-oh
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2=CC=CC=C2
371.389
371.39
≥98.0% (HPLC,T)

Chemical Identifiers

5680-86-4
371.389
TWIVXHQQTRSWGO-KRWDZBQOSA-N
3049860
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2=CC=CC=C2
C20H21NO6
MFCD00065697
z-glu obzl-oh, cbz-glu obzl-oh, 2s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, 5-benzyl n-carbobenzoxy-l-glutamate, n-cbz-l-glutamic acid 5-benzyl ester, s-2-benzyloxycarbonylamino-pentanedioic acid 5-benzyl ester, s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, z-glu obzl oh, pubchem14964, z-glu-obzl-oh
(2S)-5-oxo-5-phenylmethoxy-2-(phenylmethoxycarbonylamino)pentanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents