missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Benzotriazolecarboxylic Acid, TCI America™

$41.25 - $142.09

Chemical Identifiers

CAS 23814-12-2
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00012318
InChI Key GUOVBFFLXKJFEE-UHFFFAOYSA-N
Synonym 5-Carboxybenzotriazole
PubChem CID 72917
IUPAC Name 2H-benzotriazole-5-carboxylic acid
SMILES C1=CC2=NNN=C2C=C1C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B38695G View Documents TCI America
B38695G
5g Each for $41.25
 
B386925G View Documents TCI America
B386925G
25g Each for $142.09
 
Specifications

Chemical Identifiers

23814-12-2
163.136
GUOVBFFLXKJFEE-UHFFFAOYSA-N
72917
C1=CC2=NNN=C2C=C1C(=O)O
C7H5N3O2
MFCD00012318
5-Carboxybenzotriazole
2H-benzotriazole-5-carboxylic acid

Specifications

Undesignated
23814-12-2
MFCD00012318
GUOVBFFLXKJFEE-UHFFFAOYSA-N
2H-benzotriazole-5-carboxylic acid
72917
Crystal-Powder at 20°C
5-Benzotriazolecarboxylic Acid
5g
C7H5N3O2
5-Carboxybenzotriazole
C1=CC2=NNN=C2C=C1C(=O)O
163.136
163.14
≥96.0% (HPLC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5993&(5)-5794

TSCA : No