Learn More
5-Azaindole, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6427203
Description
5-Azaindole is a factor VIIa inhibitor. Also useful intermediate for pharmaceutical application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Azaindole is a factor VIIa inhibitor. Also useful intermediate for pharmaceutical application.
Solubility
Hardly soluble in water. Soluble in methanol and chloroform.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
5-Azaindole | |
104°C to 108°C | |
MFCD00955936 | |
109684 | |
Hardly soluble in water. Soluble in methanol and chloroform. | |
C1=CNC2=C1C=NC=C2 | |
118.139 | |
118.14 |
271-34-1 | |
C7H6N2 | |
1 g | |
5-azaindole, 1h-pyrrolo 3,2-c pyridine, 1,5-diazaindene, 3,7-dideazapurine, 5-azindole, 5h-pyrrolo 3,2-c pyridine, pyrrolo 3,2-c pyridine, 5-aza indole, 5-aza-indole | |
SRSKXJVMVSSSHB-UHFFFAOYSA-N | |
1H-pyrrolo[3,2-c]pyridine | |
9220 | |
98% |
Chemical Identifiers
271-34-1 | |
118.139 | |
SRSKXJVMVSSSHB-UHFFFAOYSA-N | |
9220 | |
C1=CNC2=C1C=NC=C2 |
C7H6N2 | |
MFCD00955936 | |
5-azaindole, 1h-pyrrolo 3,2-c pyridine, 1,5-diazaindene, 3,7-dideazapurine, 5-azindole, 5h-pyrrolo 3,2-c pyridine, pyrrolo 3,2-c pyridine, 5-aza indole, 5-aza-indole | |
1H-pyrrolo[3,2-c]pyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : UY8725000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only