missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Aza-2-deoxycytidine, +98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC396810500

 View more versions of this product

Catalog No. AC396810500


 

Specifications Description & Specifications

Specifications

5-Aza-2';-deoxycytidine
97.5
20ppm max.
1% max.
≥97.5% (HPLC)
C8H12N4O4
228.21
15, 2856
0.1% max.
50mg
2353-33-5
100.0
Authentic
Low Melting Solid
98%
Glass bottle
MFCD00043011
4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1, 3, 5-triazin-2(1H)-one
Off-white to brown

Chemical Identifiers

2353-33-5
100.0
C8H12N4O4
MFCD00043011
5-Aza-2';-deoxycytidine
97.5
98%
228.21
4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1, 3, 5-triazin-2(1H)-one
≥97.5% (HPLC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning