Please login to your online account to display your discounted pricing

5-Aza-2-deoxycytidine, +98%, ACROS Organics™


Quantity 10mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC396810100

 View more versions of this product

Catalog No. AC396810100

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Low Melting Solid
Formula Weight 228.21
Assay ≥97.5% (HPLC)
Quantity 10mg
Color Off-white to brown
Merck Index 15, 2856
CAS Max % 100.0
Loss on Drying 1% max.
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
CAS 2353-33-5
Chemical Name or Material 5-Aza-2';-deoxycytidine
Molecular Formula C8H12N4O4
Assay Percent Range 98%
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00043011
CAS Min % 97.5
Packaging Glass bottle
Sulfated Ash 0.1% max.
Synonym 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1, 3, 5-triazin-2(1H)-one
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

Specifications

Chemical Name or Material 5-Aza-2';-deoxycytidine
CAS 2353-33-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C8H12N4O4
Formula Weight 228.21
MDL Number MFCD00043011
Synonym 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1, 3, 5-triazin-2(1H)-one
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents