missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Aminouracil, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 226820025
Specifications
5-Aminouracil | |
Glass bottle | |
2.5g | |
932-52-5 | |
100.0 | |
C4H5N3O2 | |
24, 463 | |
BISHACNKZIBDFM-UHFFFAOYSA-N | |
5-amino-1H-pyrimidine-2,4-dione | |
13611 | |
127.1 | |
≥97.5% |
Authentic | |
Beige to Brown | |
98% | |
97.5 | |
98% | |
MFCD00006025 | |
2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, 5-amino-1,3-dihydropyrimidine-2,4-dione, 5-amino-2,4-dihydroxypyrimidine, 5-aminopyrimidine-2,4-diol, 5-aminouracil, r2o7eky9g9, unii-r2o7eky9g9, uracil, 5-amino, wbu | |
C1=C(C(=O)NC(=O)N1)N | |
127.103 | |
CHEBI:46348 | |
Fine Powder |
Chemical Identifiers
932-52-5 | |
127.103 | |
BISHACNKZIBDFM-UHFFFAOYSA-N | |
13611 | |
5-amino-1H-pyrimidine-2,4-dione |
C4H5N3O2 | |
MFCD00006025 | |
2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, 5-amino-1,3-dihydropyrimidine-2,4-dione, 5-amino-2,4-dihydroxypyrimidine, 5-aminopyrimidine-2,4-diol, 5-aminouracil, r2o7eky9g9, unii-r2o7eky9g9, uracil, 5-amino, wbu | |
CHEBI:46348 | |
C1=C(C(=O)NC(=O)N1)N |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
AIR SENSITIVE
EINECSNumber : 213-252-3
RTECSNumber : YQ8740000
TSCA : TSCA