missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(Aminosulfonyl)-1-methyl-1H-pyrrole-2-carboxylic acid, 97%, Maybridge

Chemical Identifiers

CAS 306936-62-9
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C6H8N2O4S
Formula Weight 204.21
MDL Number MFCD02677728
Chemical Name or Material 5-(Aminosulfonyl)-1-methyl-1H-pyrrole-2-carboxylic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
SP01488CB SDS Fisher Scientific Company
SP01488CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

306936-62-9
97%
204.21
5-(Aminosulfonyl)-1-methyl-1H-pyrrole-2-carboxylic acid
97.0
C6H8N2O4S
MFCD02677728
97%

Specifications

204.21
97.0
MFCD02677728
306936-62-9
5-(Aminosulfonyl)-1-methyl-1H-pyrrole-2-carboxylic acid
250mg
C6H8N2O4S
Amber Glass Bottle
97%
97%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.