Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Aminosalicylic acid, 95%

$74.58 - $232.04

Chemical Identifiers

Chemical Name or Material 5-Aminosalicylic acid
CAS 89-57-6
Molecular Formula C7H7NO3
Formula Weight 153.14
MDL Number MFCD00007877
Beilstein 2090421
Merck Index 14,5904
Synonym 5-Amino-2-hydroxybenzoic acid; Mesalamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2397022

  SDS
alfa aesar™
B2397022
100g Each for $74.58

AAB2397036

  SDS
alfa aesar™
B2397036
500g Each for $232.04
Description & Specifications

Specifications

Formula Weight 153.14
CAS 89-57-6
Synonym 5-Amino-2-hydroxybenzoic acid; Mesalamine
TSCA TSCA
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number VO1400000
Merck Index 14,5904
MDL Number MFCD00007877
Quantity 100g
Health Hazard 1 Warning
Assay 95%
Chemical Name or Material 5-Aminosalicylic acid
Beilstein 2090421
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈280°C (dec.)
Molecular Formula C7H7NO3

Solubility
Soluble in dimethyl sulfoxide.

Notes
Moisture, air and light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and chloroformates.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.