Learn More
5-Aminosalicylic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2397022
Description
5-Aminosalicylic acid is used in the preparation of gastrointestinal anti-inflammatory agents. It is a metabolite of sulfasalazine. It acts as a drug involved in the treatment of Crohnâs disease and ulcerative colitis. Further, it is used to make dyes and light-sensitive papers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in dimethyl sulfoxide.
Notes
Moisture, air and light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and chloroformates.
Specifications
5-Aminosalicylic acid | |
∼280°C (decomposition) | |
MFCD00007877 | |
2090421 | |
5-aminosalicylic acid, mesalazine, mesalamine, pentasa, claversal, asacol, canasa, salofalk, rowasa, 5-asa | |
KBOPZPXVLCULAV-UHFFFAOYSA-N | |
5-amino-2-hydroxybenzoic acid | |
4075 | |
153.14 |
89-57-6 | |
C7H7NO3 | |
100 g | |
14,5904 | |
Soluble in dimethyl sulfoxide. | |
C1=CC(=C(C=C1N)C(=O)O)O | |
153.137 | |
CHEBI:6775 | |
95% |
Chemical Identifiers
89-57-6 | |
153.137 | |
KBOPZPXVLCULAV-UHFFFAOYSA-N | |
4075 | |
5-amino-2-hydroxybenzoic acid |
C7H7NO3 | |
MFCD00007877 | |
5-aminosalicylic acid, mesalazine, mesalamine, pentasa, claversal, asacol, canasa, salofalk, rowasa, 5-asa | |
CHEBI:6775 | |
C1=CC(=C(C=C1N)C(=O)O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
EINECSNumber : 201-919-1
RTECSNumber : VO1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only