Learn More
5-Aminoquinoline, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 104600050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Aminoquinoline | |
106.0°C to 109.0°C | |
310.0°C | |
Glass bottle | |
MFCD00006797 | |
22, 445 | |
XMIAFAKRAAMSGX-UHFFFAOYSA-N | |
quinolin-5-amine | |
11911 | |
≥96.0% (GC) |
611-34-7 | |
White to Yellow | |
Authentic | |
C9H8N2 | |
5 g | |
5-aminoquinoline, 5-quinolinamine, quinoline, 5-amino, 5-quinolylamine, quinolin-5-ylamine, unii-yyj34z13sh, 5-amino-quinoline, ccris 1680, yyj34z13sh | |
NC1=C2C=CC=NC2=CC=C1 | |
144.18 | |
144.18 | |
Crystalline Powder or Granules |
Chemical Identifiers
611-34-7 | |
144.18 | |
XMIAFAKRAAMSGX-UHFFFAOYSA-N | |
11911 | |
NC1=C2C=CC=NC2=CC=C1 |
C9H8N2 | |
MFCD00006797 | |
5-aminoquinoline, 5-quinolinamine, quinoline, 5-amino, 5-quinolylamine, quinolin-5-ylamine, unii-yyj34z13sh, 5-amino-quinoline, ccris 1680, yyj34z13sh | |
quinolin-5-amine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 210-266-1
RUO – Research Use Only