missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Aminopyrazine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H66207MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Aminopyrazine-2-carboxylic acid | |
C5H5N3O2 | |
250 mg | |
5-amino-2-pyrazinecarboxylic acid, 2-amino-5-pyrazinecarboxylic acid, pyrazinecarboxylic acid, 5-amino, 5-amino-pyrazine-2-carboxylic acid, 5-aminopyrazine-2-carboxylicacid, 5-amino-pyrazine-2-carboxylicacid, pubchem7866, 5-amino-pyrazinecarboxylic acid, 2-amino-5-pirazinecarboxylic acid | |
C1=C(N=CC(=N1)N)C(=O)O | |
139.114 | |
139.11 | |
Solid |
40155-43-9 | |
MFCD07371664 | |
120673 | |
FMEFOOOBIHQRMK-UHFFFAOYSA-N | |
5-aminopyrazine-2-carboxylic acid | |
21897334 | |
95% |
Chemical Identifiers
40155-43-9 | |
139.114 | |
FMEFOOOBIHQRMK-UHFFFAOYSA-N | |
21897334 | |
C1=C(N=CC(=N1)N)C(=O)O |
C5H5N3O2 | |
MFCD07371664 | |
5-amino-2-pyrazinecarboxylic acid, 2-amino-5-pyrazinecarboxylic acid, pyrazinecarboxylic acid, 5-amino, 5-amino-pyrazine-2-carboxylic acid, 5-aminopyrazine-2-carboxylicacid, 5-amino-pyrazine-2-carboxylicacid, pubchem7866, 5-amino-pyrazinecarboxylic acid, 2-amino-5-pirazinecarboxylic acid | |
5-aminopyrazine-2-carboxylic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only