Please login to your online account to display your discounted pricing

5-Aminonicotinic acid, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Aminonicotinic acid
CAS 24242-19-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H6N2O2
Formula Weight 138.13
MDL Number MFCD00129116
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361140050

  SDS
thermo fisher scientific
AC361140050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 138.13
Quantity 5g
Color Golden-brown
CAS Min % 94.0
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 5-Aminonicotinic acid
Assay Percent Range 95%
Molecular Formula C6H6N2O2
MDL Number MFCD00129116
CAS 24242-19-1
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.