Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Aminomethyl-2-(Boc-amino)pyridine, 97%

$269.72 - $1,193.35

Chemical Identifiers

Chemical Name or Material 5-Aminomethyl-2-(Boc-amino)pyridine
CAS 187237-37-2
Molecular Formula C11H17N3O2
Formula Weight 223.28
MDL Number MFCD07781132
Synonym tert-Butyl (5-aminomethyl-2-pyridyl)carbamate; 2-(Boc-amino)-5-(aminomethyl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5007803

  SDS
alfa aesar™
H5007803
1g Each for $269.72

AAH5007806

  SDS
alfa aesar™
H5007806
5g Each for $1,193.35
Description & Specifications

Specifications

Formula Weight 223.28
Chemical Name or Material 5-Aminomethyl-2-(Boc-amino)pyridine
Assay 97%
Molecular Formula C11H17N3O2
Melting Point 132° to 134°C
MDL Number MFCD07781132
Quantity 1g
CAS 187237-37-2
Synonym tert-Butyl (5-aminomethyl-2-pyridyl)carbamate; 2-(Boc-amino)-5-(aminomethyl)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.