Learn More
5-Aminoisoquinoline, 99%, Thermo Scientific Chemicals
$73.25 - $676.71
Chemical Identifiers
CAS | 1125-60-6 |
---|---|
Molecular Formula | C9H8N2 |
Molecular Weight (g/mol) | 144.18 |
MDL Number | MFCD00006907 |
InChI Key | DTVYNUOOZIKEEX-UHFFFAOYSA-N |
Synonym | 5-aminoisoquinoline, 5-isoquinolinamine, isoquinoline, 5-amino, isoquinol-5-ylamine, isoquinolin-5-ylamine, 5-amino-isoquinoline, 5-aminoisoquinolin, 5-isoquinolylamine, 5iq, 5aminoisoquinoline |
PubChem CID | 70766 |
IUPAC Name | isoquinolin-5-amine |
SMILES | NC1=C2C=CN=CC2=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0122303
|
Thermo Scientific Chemicals
L0122303 |
1 g |
Each for $73.25
|
|
|||||
AAL0122306
|
Thermo Scientific Chemicals
L0122306 |
5 g |
Each for $225.03
|
|
|||||
AAL0122314
|
Thermo Scientific Chemicals
L0122314 |
25 g |
Each for $676.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1125-60-6 | |
144.18 | |
DTVYNUOOZIKEEX-UHFFFAOYSA-N | |
70766 | |
NC1=C2C=CN=CC2=CC=C1 |
C9H8N2 | |
MFCD00006907 | |
5-aminoisoquinoline, 5-isoquinolinamine, isoquinoline, 5-amino, isoquinol-5-ylamine, isoquinolin-5-ylamine, 5-amino-isoquinoline, 5-aminoisoquinolin, 5-isoquinolylamine, 5iq, 5aminoisoquinoline | |
isoquinolin-5-amine |
Specifications
1125-60-6 | |
C9H8N2 | |
1 g | |
Light sensitive | |
DTVYNUOOZIKEEX-UHFFFAOYSA-N | |
isoquinolin-5-amine | |
70766 | |
99% |
126°C to 131°C | |
MFCD00006907 | |
114465 | |
5-aminoisoquinoline, 5-isoquinolinamine, isoquinoline, 5-amino, isoquinol-5-ylamine, isoquinolin-5-ylamine, 5-amino-isoquinoline, 5-aminoisoquinolin, 5-isoquinolylamine, 5iq, 5aminoisoquinoline | |
NC1=C2C=CN=CC2=CC=C1 | |
144.18 | |
144.18 | |
5-Aminoisoquinoline |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 214-408-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only