missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Aminoindole, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 103900050
Specifications
5-Aminoindole | |
Glass bottle | |
130.0°C to 134.0°C | |
97% | |
96.0 | |
96% min. (HPLC) | |
MFCD00005679 | |
Solubility in water: insoluble. Other solubilities: slightly soluble in most common organic solvents | |
C1=CC2=C(C=CN2)C=C1N | |
132.16 | |
CHEBI:33067 | |
Crystalline Powder |
Authentic | |
Brown-Gray to White | |
5g | |
5192-03-0 | |
100.0 | |
C8H8N2 | |
1h-indol-5-ylamine, 5-amino indole, 5-amino-indole, 5-aminoindole, 5-indolamine, indol-5-ylamine, indole, 5-amino, indole-5-ylamine, unii-q732pg0y16 | |
ZCBIFHNDZBSCEP-UHFFFAOYSA-N | |
1H-indol-5-amine | |
78867 | |
132.16 | |
97% |
Chemical Identifiers
5192-03-0 | |
132.16 | |
ZCBIFHNDZBSCEP-UHFFFAOYSA-N | |
78867 | |
1H-indol-5-amine |
C8H8N2 | |
MFCD00005679 | |
1h-indol-5-ylamine, 5-amino indole, 5-amino-indole, 5-aminoindole, 5-indolamine, indol-5-ylamine, indole, 5-amino, indole-5-ylamine, unii-q732pg0y16 | |
CHEBI:33067 | |
C1=CC2=C(C=CN2)C=C1N |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
GHS Signal Word: Warning
EINECSNumber : 225-977-2
TSCA : TSCA
Recommended Storage : −20°C