Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Aminoindole, 97%

$113.76 - $1,396.12

Chemical Identifiers

Chemical Name or Material 5-Aminoindole
CAS 5192-03-0
Molecular Formula C8H8N2
Formula Weight 132.17
MDL Number MFCD00005679
Beilstein 112348
Synonym 5-Indolamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2439103

  SDS
alfa aesar™
B2439103
1g Each for $113.76

AAB2439106

  SDS
alfa aesar™
B2439106
5g Each for $352.50

AAB2439114

  SDS
alfa aesar™
B2439114
25g Each for $1,396.12
Description & Specifications

Specifications

Formula Weight 132.17
Chemical Name or Material 5-Aminoindole
TSCA TSCA
Assay 97%
Molecular Formula C8H8N2
Melting Point 130° to 134°C
MDL Number MFCD00005679
CAS 5192-03-0
Quantity 1g
Beilstein 112348
Synonym 5-Indolamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C