Learn More
5-Aminoindole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2439106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Aminoindole | |
130°C to 134°C | |
MFCD00005679 | |
112348 | |
5-aminoindole, indol-5-ylamine, indole, 5-amino, unii-q732pg0y16, 1h-indol-5-ylamine, indole-5-ylamine, 5-indolamine, 5-amino indole, 5-amino-indole | |
NC1=CC=C2NC=CC2=C1 | |
132.17 | |
CHEBI:33067 | |
97% |
5192-03-0 | |
C8H8N2 | |
5 g | |
Air sensitive | |
ZCBIFHNDZBSCEP-UHFFFAOYSA-N | |
1H-indol-5-amine | |
78867 | |
132.17 |
Chemical Identifiers
5192-03-0 | |
132.17 | |
ZCBIFHNDZBSCEP-UHFFFAOYSA-N | |
78867 | |
1H-indol-5-amine |
C8H8N2 | |
MFCD00005679 | |
5-aminoindole, indol-5-ylamine, indole, 5-amino, unii-q732pg0y16, 1h-indol-5-ylamine, indole-5-ylamine, 5-indolamine, 5-amino indole, 5-amino-indole | |
CHEBI:33067 | |
NC1=CC=C2NC=CC2=C1 |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 225-977-2
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only