missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Aminoimidazole-4-carboxamide, 95%, ACROS Organics™

$129.87 - $129.87

Chemical Identifiers

CAS 360-97-4
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C4H6N4O
Formula Weight 126.12
MDL Number MFCD02181040
Chemical Name or Material 5-Aminoimidazole-4-carboxamide
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC397550250 SDS Thermo Fisher Scientific
AC397550250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC397550050 SDS Thermo Fisher Scientific
AC397550050
Glass bottle 5g Each for $129.87
Specifications Description & Specifications

Chemical Identifiers

360-97-4
100.0
C4H6N4O
MFCD02181040
≥94%
94.0
95%
126.12
5-Aminoimidazole-4-carboxamide

Specifications

360-97-4
100.0
Glass bottle
95%
166° to 170°C
MFCD02181040
25g
≥94%
94.0
Authentic
Crystalline Powder
C4H6N4O
126.12
Gray to purple
5-Aminoimidazole-4-carboxamide

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.