Please login to your online account to display your discounted pricing

5-Aminobenzene-1,3-diol hydrochloride, Technical Grade, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Aminobenzene-1, 3-diol hydrochloride
Assay Tech.
Molecular Formula C6H7NO2·ClH
Linear Formula NH2C6H3(OH)2.HCl
Formula Weight 161.59
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB10193DE

 
fisher scientific company
BTB10193DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB10193DA

  SDS
fisher scientific company
BTB10193DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB10193CB

  SDS
fisher scientific company
BTB10193CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Technical Grade
Formula Weight 161.59
Quantity 5g
Assay Tech.
Packaging Amber Glass Bottle
Chemical Name or Material 5-Aminobenzene-1, 3-diol hydrochloride
Linear Formula NH2C6H3(OH)2.HCl
CAS 6318-56-5
Molecular Formula C6H7NO2·ClH