Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-7-azaindole, 97%

Chemical Identifiers

Chemical Name or Material 5-Amino-7-azaindole
CAS 100960-07-4
Formula Weight 133.15
MDL Number MFCD06659683
Synonym 5-Amino-1H-pyrrolo[2,3-b]pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3538203

  SDS
alfa aesar™
H3538203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH35382MD

  SDS
alfa aesar™
H35382MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 129° to 131°C
Formula Weight 133.15
Chemical Name or Material 5-Amino-7-azaindole
Physical Form Solid
Assay 97%
MDL Number MFCD06659683
CAS 100960-07-4
Color Yellow to yellow/brown
Synonym 5-Amino-1H-pyrrolo[2,3-b]pyridine
Quantity 1g
Sensitivity Light Sensitive
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.