Please login to your online account to display your discounted pricing

5-Amino-6-nitroquinoline, 98%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  163300010

 View more versions of this product

Catalog No. AC163300010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 189.17
Physical Form Fluffy Powder
Quantity 1g
Color Ochre-yellow
EINECS Number 252-822-6
CAS Max % 100.0
MDL Number MFCD00006798
Infrared Spectrum Authentic
Chemical Name or Material 5-Amino-6-nitroquinoline
CAS 35975-00-9
Assay Percent Range 98%
CAS Min % 97.5
Molecular Formula C9 H7 N3 O2
Name Note 98%
Packaging Glass bottle

Specifications

Chemical Name or Material 5-Amino-6-nitroquinoline
Name Note 98%
CAS 35975-00-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H7 N3 O2
Formula Weight 189.17
MDL Number MFCD00006798