Please login to your online account to display your discounted pricing

5-Amino-6-nitroquinoline, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Amino-6-nitroquinoline
Name Note 98%
CAS 35975-00-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H7 N3 O2
Formula Weight 189.17
MDL Number MFCD00006798
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC163300010

  SDS
thermo fisher scientific
163300010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fluffy Powder
Assay ≥97.5%
Formula Weight 189.17
Quantity 1g
Color Ochre-yellow
EINECS Number 252-822-6
CAS Max % 100.0
Chemical Name or Material 5-Amino-6-nitroquinoline
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9 H7 N3 O2
MDL Number MFCD00006798
CAS 35975-00-9
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle